Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stearic acid-d3 | 62163-39-7 | MFCD00145327 | 99.9% | 287.50 g/mol | C18H33D3O2 | 10 MG
SDP

Catalog No. 5000297311
Encompass_Preferred

Stearic acid-d3 is the deuterium-labeled form of stearic acid, a long-chain saturated fatty acid commonly found in animal and vegetable fats and oils. It is supplied as a high-purity labeled standard for research and analytical applications, and is intended for use as a tracer and internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity: 99.9%.
  • Molecular weight: 287.50 g/mol.
  • Chemical formula: C18H33D3O2.
  • Available in small pack sizes suitable for analytical workflows.

Catalog No. 50-002-97311 Supplier Medchemexpress LLC Supplier No. HYB2219S310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stearic acid-d3 is the deuterium-labeled form of stearic acid, a long-chain saturated fatty acid commonly found in animal and vegetable fats and oils. It is supplied as a high-purity labeled standard for research and analytical applications, and is intended for use as a tracer and internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity: 99.9%.
  • Molecular weight: 287.50 g/mol.
  • Chemical formula: C18H33D3O2.
  • Available in small pack sizes suitable for analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.