Learn More
Cayman Chemical Stearic Acid-d35, Purity: ≥99% deuterated forms (d1-d35), CAS: 17660-51-4, 250 mg
SDP
Catalog No. 502894410
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-289-4410 Supplier Cayman Chemical Supplier No. 9003318250 MG
An internal standard for the quantification of stearic acid by GC- or LC-MS