Learn More
Cayman Chemical StearIc AcId-d35 50mg
SDP
Catalog No. 502892682
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-289-2682 Supplier Cayman Chemical Supplier No. 900331850 MG
An internal standard for the quantification of stearic acid by GC- or LC-MS