Learn More
Cayman Chemical Stearic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 130048-57-6, 10 mg
SDP
Catalog No. 502992417
Encompass_Preferred
An internal standard for the quantification of stearicacid by GC- or LC-MS
Catalog No. 50-299-2417 Supplier Cayman Chemical Supplier No. 3054910 MG
An internal standard for the quantification of stearicacid by GC- or LC-MS