Learn More
Cayman Chemical Stearic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 130048-57-6, 250 mg
SDP
Catalog No. 502990903
Encompass_Preferred
An internal standard for the quantification of stearicacid by GC- or LC-MS
Catalog No. 50-299-0903 Supplier Cayman Chemical Supplier No. 30549250 MG
An internal standard for the quantification of stearicacid by GC- or LC-MS