Learn More
Cayman Chemical StearIc AcId-d4 50mg
SDP
Catalog No. 502992418
Encompass_Preferred
An internal standard for the quantification of stearicacid by GC- or LC-MS
Catalog No. 50-299-2418 Supplier Cayman Chemical Supplier No. 3054950 MG
An internal standard for the quantification of stearicacid by GC- or LC-MS