Learn More
Cayman Chemical Stearic Acid-d5, Purity: ≥99% deuterated forms (d1-d5), CAS: 211443-83-3, 10 mg
SDP
Catalog No. 502895197
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-289-5197 Supplier Cayman Chemical Supplier No. 900045510 MG
An internal standard for the quantification of stearic acid by GC- or LC-MS