Learn More
Cayman Chemical StearIc AcId-d5 1mg
SDP
Catalog No. 502892562
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-289-2562 Supplier Cayman Chemical Supplier No. 90004551 MG
Add to Cart
Add to Cart
An internal standard for the quantification of stearic acid by GC- or LC-MS