Learn More
Medchemexpress LLC Stearic acid-d5 | 211443-83-3 | 5 MG
SDP
Catalog No. 5000409684
Encompass_Preferred
Stearic acid-d5 is a deuterium labeled stearic acid, which is a long-chain dietary saturated fatty acid found in many animal and vegetable fats and oils. It is used as a tracer for quantitation in drug development, with deuteration potentially affecting pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled stearic acid
  • Used as a tracer for quantitation in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
Catalog No. 50-004-09684 Supplier Medchemexpress LLC Supplier No. HYB2219S65MG
Stearic acid-d5 is a deuterium labeled stearic acid, which is a long-chain dietary saturated fatty acid found in many animal and vegetable fats and oils. It is used as a tracer for quantitation in drug development, with deuteration potentially affecting pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled stearic acid
  • Used as a tracer for quantitation in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month