Learn More
Medchemexpress LLC Stearic acid-d5 | 211443-83-3 | 50 MG
SDP
Catalog No. 5000410386
Encompass_Preferred
Stearic acid-d5 is the deuterium labeled Stearic acid. Stearic acid is a long chain dietary saturated fatty acid which exists in many animal and vegetable fats and oils.
  • Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation.
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-10386 Supplier Medchemexpress LLC Supplier No. HYB2219S650MG
Stearic acid-d5 is the deuterium labeled Stearic acid. Stearic acid is a long chain dietary saturated fatty acid which exists in many animal and vegetable fats and oils.
  • Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation.
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs.