Learn More
Medchemexpress LLC Stearic acid-d5 | 211443-83-3 | ≥99% | 100 MG
SDP
Catalog No. 5000410540
Encompass_Preferred
Stearic acid-d5 is a deuterium labeled stearic acid, a long-chain dietary saturated fatty acid found in various animal and vegetable fats and oils. It is commonly used as an internal standard for the quantification of stearic acid by GC- or LC-MS. Deuterium labeling incorporates stable heavy isotopes into molecules, which can serve as tracers for quantitation during drug development and may impact pharmacokinetic and metabolic profiles of drugs.
  • Used as an internal standard for quantification.
  • Deuterium labeled.
  • Functions as a tracer for quantitation during drug development.
  • May affect pharmacokinetic and metabolic profiles.
  • Long-chain saturated fatty acid.
Catalog No. 50-004-10540 Supplier Medchemexpress LLC Supplier No. HYB2219S6100MG
Stearic acid-d5 is a deuterium labeled stearic acid, a long-chain dietary saturated fatty acid found in various animal and vegetable fats and oils. It is commonly used as an internal standard for the quantification of stearic acid by GC- or LC-MS. Deuterium labeling incorporates stable heavy isotopes into molecules, which can serve as tracers for quantitation during drug development and may impact pharmacokinetic and metabolic profiles of drugs.
  • Used as an internal standard for quantification.
  • Deuterium labeled.
  • Functions as a tracer for quantitation during drug development.
  • May affect pharmacokinetic and metabolic profiles.
  • Long-chain saturated fatty acid.