Learn More
Cayman Chemical StearIc AcId-d5 25mg
SDP
Catalog No. 502892563
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-289-2563 Supplier Cayman Chemical Supplier No. 900045525 MG
An internal standard for the quantification of stearic acid by GC- or LC-MS