Learn More
Cayman Chemical Stearic Acid-d5, Purity: ≥99% deuterated forms (d1-d5), CAS: 211443-83-3, 5 mg
SDP
Catalog No. 502892564
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-289-2564 Supplier Cayman Chemical Supplier No. 90004555 MG
An internal standard for the quantification of stearic acid by GC- or LC-MS