Learn More
Medchemexpress LLC Stearic acid-d7 | 951209-59-9 | 98.0% | 1 MG
SDP
Catalog No. 5000410387
Encompass_Preferred
Stearic acid-d7 is the deuterium labeled Stearic acid. Stearic acid is a long chain dietary saturated fatty acid which exists in many animal and vegetable fats and oils.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-10387 Supplier Medchemexpress LLC Supplier No. HYB2219S91MG
Stearic acid-d7 is the deuterium labeled Stearic acid. Stearic acid is a long chain dietary saturated fatty acid which exists in many animal and vegetable fats and oils.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.