Learn More
Medchemexpress LLC Stearic acid-d7 | 951209-59-9 | 98.0% | 25 MG
SDP
Catalog No. 5000411969
Encompass_Preferred
Stearic acid-d7 is a deuterium-labeled stearic acid, a long-chain dietary saturated fatty acid found in many animal and vegetable fats and oils.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11969 Supplier Medchemexpress LLC Supplier No. 25MGHYB2219S9
Stearic acid-d7 is a deuterium-labeled stearic acid, a long-chain dietary saturated fatty acid found in many animal and vegetable fats and oils.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes can affect pharmacokinetic and metabolic profiles of drugs.