Learn More
Medchemexpress LLC Stearic acid-d7 | 951209-59-9 | 98.0% | 5 MG
SDP
Catalog No. 5000410637
Encompass_Preferred
Stearic acid-d7 is the deuterium labeled Stearic acid. Stearic acid is a long chain dietary saturated fatty acid which exists in many animal and vegetable fats and oils.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-10637 Supplier Medchemexpress LLC Supplier No. HYB2219S95MG
Stearic acid-d7 is the deuterium labeled Stearic acid. Stearic acid is a long chain dietary saturated fatty acid which exists in many animal and vegetable fats and oils.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.