Learn More
Cayman Chemical Stearic Acid Fatty Acids and D
SDP
Catalog No. 502887921
Encompass_Preferred
A long-chain saturated fatty acid; a major component of cocoa butter and has also been found in beef fat and vegetable oils; does not induce hypercholesterolemia or raise LDL-cholesterol unlike many long-chain saturated fatty acids
Catalog No. 50-288-7921 Supplier Cayman Chemical Supplier No. 1001129810 G
A long-chain saturated fatty acid; a major component of cocoa butter and has also been found in beef fat and vegetable oils; does not induce hypercholesterolemia or raise LDL-cholesterol unlike many long-chain saturated fatty acids