Learn More
Cayman Chemical Stearic Acid, Purity: ≥98%, CAS: 57-11-4, 500 mg
SDP
Catalog No. 502890660
Encompass_Preferred
A long-chain saturated fatty acid; a major component of cocoa butter and has also been found in beef fat and vegetable oils; does not induce hypercholesterolemia or raise LDL-cholesterol unlike many long-chain saturated fatty acids
Catalog No. 50-289-0660 Supplier Cayman Chemical Supplier No. 10011298500 MG
A long-chain saturated fatty acid; a major component of cocoa butter and has also been found in beef fat and vegetable oils; does not induce hypercholesterolemia or raise LDL-cholesterol unlike many long-chain saturated fatty acids