Learn More
Cayman Chemical Stearic Acid Fatty Acids and D
SDP
Supplier:  Cayman Chemical 100112985 G
Encompass_Preferred
A long-chain saturated fatty acid; a major component of cocoa butter and has also been found in beef fat and vegetable oils; does not induce hypercholesterolemia or raise LDL-cholesterol unlike many long-chain saturated fatty acids
Catalog No. 50-288-7811