Learn More
Cayman Chemical StearIc AcId methyl ester 1g
SDP
Catalog No. 502984026
Encompass_Preferred
An esterified version of the free acid which is less water soluble but more amenable for the formulation of stearate-containing diets and dietary supplements
Catalog No. 50-298-4026 Supplier Cayman Chemical Supplier No. 206091 G
An esterified version of the free acid which is less water soluble but more amenable for the formulation of stearate-containing diets and dietary supplements