Learn More
Cayman Chemical Stearic Acid methyl ester, Purity: ≥95%, CAS: 112-61-8, 5 g
SDP
Catalog No. 502984027
Encompass_Preferred
An esterified version of the free acid which is less water soluble but more amenable for the formulation of stearate-containing diets and dietary supplements
Catalog No. 50-298-4027 Supplier Cayman Chemical Supplier No. 206095 G
An esterified version of the free acid which is less water soluble but more amenable for the formulation of stearate-containing diets and dietary supplements