Learn More
Cayman Chemical Stearic Acid methyl ester, Purity: ≥95%, CAS: 112-61-8, 500 mg
SDP
Catalog No. 502982067
Encompass_Preferred
An esterified version of the free acid which is less water soluble but more amenable for the formulation of stearate-containing diets and dietary supplements
Catalog No. 50-298-2067 Supplier Cayman Chemical Supplier No. 20609500 MG
An esterified version of the free acid which is less water soluble but more amenable for the formulation of stearate-containing diets and dietary supplements