Learn More
Medchemexpress LLC Stearic acid-mPEG | 99.64% | 2000 | 500 MG
SDP
Catalog No. 5000391154
Encompass_Preferred
Stearic acid-mPEG is a PEG lipid designed to improve the delivery efficiency and tissue specificity of poorly soluble drugs. It is suitable for drug delivery research.
  • Improves delivery efficiency of poorly soluble drugs
  • Enhances tissue specificity
  • Useful in drug delivery research
Catalog No. 50-003-91154 Supplier Medchemexpress LLC Supplier No. HY166979500MG
Stearic acid-mPEG is a PEG lipid designed to improve the delivery efficiency and tissue specificity of poorly soluble drugs. It is suitable for drug delivery research.
  • Improves delivery efficiency of poorly soluble drugs
  • Enhances tissue specificity
  • Useful in drug delivery research