Learn More
Medchemexpress LLC Stearic acid-mPEG (MW 2000) | 99.6% | 2000 (Average) | 50 MG
SDP
Catalog No. 5000390692
Encompass_Preferred
Stearic acid-mPEG is a polyethylene glycol (PEG) lipid designed to enhance the delivery efficiency and tissue specificity of poorly soluble drugs. This compound is valuable for researchers in drug delivery, offering improved drug targeting and absorption.
  • Enhances targeting of poorly soluble compounds.
  • Improves absorption in specific tissues.
  • Supports advanced drug delivery research.
Catalog No. 50-003-90692 Supplier Medchemexpress LLC Supplier No. HY16697950MG
Stearic acid-mPEG is a polyethylene glycol (PEG) lipid designed to enhance the delivery efficiency and tissue specificity of poorly soluble drugs. This compound is valuable for researchers in drug delivery, offering improved drug targeting and absorption.
  • Enhances targeting of poorly soluble compounds.
  • Improves absorption in specific tissues.
  • Supports advanced drug delivery research.