Learn More
Medchemexpress LLC Stearic acid-PEG-Mal | 5000 (Average) | 250 MG
SDP
Catalog No. 5000281259
Encompass_Preferred
Stearic acid-PEG-Mal (average MW 5000) is a heterobifunctional pegylated lipid consisting of a stearic acid moiety linked to a polyethylene glycol chain terminated with a maleimide group. It is used as a thiol-reactive amphiphilic reagent for conjugation to cysteine-containing biomolecules, surface modification, and nanoparticle functionalization in biochemical assays. For research use only.
  • Heterobifunctional reagent with maleimide for thiol conjugation.
  • Average molecular weight 5000 providing a PEG spacer.
  • Amphiphilic stearic acid tail enables membrane or particle incorporation.
  • Suitable for surface modification and nanoparticle functionalization.
  • Storage: sealed at 4°C; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
Catalog No. 50-002-81259 Supplier Medchemexpress LLC Supplier No. HYW440952250MG
Stearic acid-PEG-Mal (average MW 5000) is a heterobifunctional pegylated lipid consisting of a stearic acid moiety linked to a polyethylene glycol chain terminated with a maleimide group. It is used as a thiol-reactive amphiphilic reagent for conjugation to cysteine-containing biomolecules, surface modification, and nanoparticle functionalization in biochemical assays. For research use only.
  • Heterobifunctional reagent with maleimide for thiol conjugation.
  • Average molecular weight 5000 providing a PEG spacer.
  • Amphiphilic stearic acid tail enables membrane or particle incorporation.
  • Suitable for surface modification and nanoparticle functionalization.
  • Storage: sealed at 4°C; in solvent store at -80°C up to 6 months or -20°C up to 1 month.