Learn More
Medchemexpress LLC Stearic acid-PEG-Mal | 5000 Da (average) | 100 MG
SDP
Catalog No. 5000281258
Encompass_Preferred
Stearic acid-PEG-Mal is a maleimide-terminated pegylated lipid (average MW 5,000 Da) used as a reagent for thiol-selective conjugation and surface functionalization. The material combines a hydrophobic stearic acid tail with a polyethylene glycol linker to aid micelle and nanoparticle formation and provide steric stabilization in formulations.
  • Maleimide-terminated PEG enables thiol-selective bioconjugation.
  • Stearic acid hydrophobic tail supports micelle and nanoparticle formation.
  • Average molecular weight 5,000 Da provides steric stabilization.
  • Solid, white to off-white reagent grade material for research use.
  • Store sealed at 4°C; in solvent: -80°C up to 6 months or -20°C up to 1 month.
Catalog No. 50-002-81258 Supplier Medchemexpress LLC Supplier No. HYW440952100MG
Stearic acid-PEG-Mal is a maleimide-terminated pegylated lipid (average MW 5,000 Da) used as a reagent for thiol-selective conjugation and surface functionalization. The material combines a hydrophobic stearic acid tail with a polyethylene glycol linker to aid micelle and nanoparticle formation and provide steric stabilization in formulations.
  • Maleimide-terminated PEG enables thiol-selective bioconjugation.
  • Stearic acid hydrophobic tail supports micelle and nanoparticle formation.
  • Average molecular weight 5,000 Da provides steric stabilization.
  • Solid, white to off-white reagent grade material for research use.
  • Store sealed at 4°C; in solvent: -80°C up to 6 months or -20°C up to 1 month.