Learn More
Medchemexpress LLC Stearic acid-PEG-NHS | 2000 Da (average) | 5 MG
SDP
Catalog No. 5000280987
Encompass_Preferred
Stearic acid-PEG-NHS (MW 2000) is a pegylated stearyl lipid bearing an N-hydroxysuccinimide (NHS) ester terminal group. The amphiphilic polymer self-assembles in aqueous media to form micelles or lipid bilayers and is used as a reactive building block for nanoparticle and liposome formulation, surface PEGylation, and bioconjugation via primary amines. Supplied reagent grade for research use only.
  • Amphiphilic pegylated lipid that self-assembles in aqueous media.
  • Terminal NHS ester reactive with primary amines to form stable amide bonds.
  • Average molecular weight approximately 2000 Da.
  • Reagent-grade material intended for research use.
  • Suitable for nanoparticle and liposome formulation and surface functionalization.
Catalog No. 50-002-80987 Supplier Medchemexpress LLC Supplier No. HYW4409275MG
Stearic acid-PEG-NHS (MW 2000) is a pegylated stearyl lipid bearing an N-hydroxysuccinimide (NHS) ester terminal group. The amphiphilic polymer self-assembles in aqueous media to form micelles or lipid bilayers and is used as a reactive building block for nanoparticle and liposome formulation, surface PEGylation, and bioconjugation via primary amines. Supplied reagent grade for research use only.
  • Amphiphilic pegylated lipid that self-assembles in aqueous media.
  • Terminal NHS ester reactive with primary amines to form stable amide bonds.
  • Average molecular weight approximately 2000 Da.
  • Reagent-grade material intended for research use.
  • Suitable for nanoparticle and liposome formulation and surface functionalization.