Learn More
Medchemexpress LLC Stearic acid-PEG-NHS | 90.0% | 2000 (average) | 10 MG
SDP
Catalog No. 5000281128
Encompass_Preferred
Stearic acid-PEG-NHS (average MW 2000) is an amphiphilic PEGylated lipid bearing an N-hydroxysuccinimide (NHS) ester. It is used as a reactive lipid for covalent conjugation to primary amines and for formulation of micelles, liposomes, and PEGylated nanoparticles in research applications.
  • Reactive NHS ester for amine conjugation.
  • Amphiphilic PEGylated lipid suitable for micelle and liposome formation.
  • Average molecular weight 2000 provides defined PEG chain length.
  • Specified reagent-grade purity for research use.
  • Recommended storage under nitrogen at low temperature to preserve activity.
Catalog No. 50-002-81128 Supplier Medchemexpress LLC Supplier No. HYW44092710MG
Stearic acid-PEG-NHS (average MW 2000) is an amphiphilic PEGylated lipid bearing an N-hydroxysuccinimide (NHS) ester. It is used as a reactive lipid for covalent conjugation to primary amines and for formulation of micelles, liposomes, and PEGylated nanoparticles in research applications.
  • Reactive NHS ester for amine conjugation.
  • Amphiphilic PEGylated lipid suitable for micelle and liposome formation.
  • Average molecular weight 2000 provides defined PEG chain length.
  • Specified reagent-grade purity for research use.
  • Recommended storage under nitrogen at low temperature to preserve activity.