Learn More
Medchemexpress LLC Stearic acid (Standard) | 57-11-4 | 99.9% | 100 MG
SDP
Catalog No. 5000409656
Encompass_Preferred
Stearic acid (Standard) is the analytical standard of Stearic acid, intended for research and analytical applications. It is a long-chain dietary saturated fatty acid that can significantly reduce visceral fat by inducing apoptosis of preadipocytes. Stearic acid can be used in the study of cardiovascular and metabolic diseases. This compound is the grade of analytical standard, which is the reference standard supplied for assay and commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Reduces visceral fat by inducing apoptosis of preadipocytes
  • Used in cardiovascular and metabolic diseases studies
  • Reference standard for assays
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-004-09656 Supplier Medchemexpress LLC Supplier No. HYB2219R100MG
Stearic acid (Standard) is the analytical standard of Stearic acid, intended for research and analytical applications. It is a long-chain dietary saturated fatty acid that can significantly reduce visceral fat by inducing apoptosis of preadipocytes. Stearic acid can be used in the study of cardiovascular and metabolic diseases. This compound is the grade of analytical standard, which is the reference standard supplied for assay and commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Reduces visceral fat by inducing apoptosis of preadipocytes
  • Used in cardiovascular and metabolic diseases studies
  • Reference standard for assays
  • Suitable for HPLC, GC, and MS experiments