Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearic Acid, Type 50, Powder, NF, 40-60%, Spectrum™ Chemical
SDP

Catalog No. 18607010 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-010 500 g Poly Pail
18-607-011 2.5 kg Poly Pail
18-607-012 12 kg Poly Pail
18-607-013 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-010 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ST110500GM
Only null left
Add to Cart
Add to Cart

ST110, 57-11-4

Spectrum™ Chemical Stearic Acid, Type 50, Powder, NF is used as an emulsifying agent, solubilizing agent, tablet and capsule lubricant. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-11-4 , 57-10-3
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.48
MDL Number MFCD00002752
InChI Key QIQXTHQIDYTFRH-UHFFFAOYSA-N
IUPAC Name octadecanoic acid
SMILES CCCCCCCCCCCCCCCCCC(O)=O

Specifications

CAS Max % 1
Boiling Point 360°C
Ignition Residue 0.001
Packaging Poly Pail
Quantity 500 g
Percent Purity 40 to 60%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Stearic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.