Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stearic anhydride | 638-08-4 | MFCD00009004 | 99.2% | 550.94 | C36H70O3 | 5 G
SDP

Catalog No. 5000282686
Encompass_Preferred

Stearic anhydride is an organic anhydride used as a biochemical reagent in life-science research. Supplied as a white to off-white solid, it is used as a synthetic intermediate and reagent in biochemical assays and organic synthesis. Intended for research use only; not for human or clinical use.

  • White to off-white solid appearance.
  • High purity (99.24%).
  • Molecular formula C36H70O3; molecular weight 550.94 g·mol-1.
  • CAS number 638-08-4.
  • Available in small laboratory pack sizes (5 g, 10 g, 50 g).
  • Store at room temperature; in solution: -80 °C (2 years), -20 °C (1 year).
  • For research use only; not for sale to patients.

Catalog No. 50-002-82686 Supplier Medchemexpress LLC Supplier No. HYW1275475G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stearic anhydride is an organic anhydride used as a biochemical reagent in life-science research. Supplied as a white to off-white solid, it is used as a synthetic intermediate and reagent in biochemical assays and organic synthesis. Intended for research use only; not for human or clinical use.

  • White to off-white solid appearance.
  • High purity (99.24%).
  • Molecular formula C36H70O3; molecular weight 550.94 g·mol-1.
  • CAS number 638-08-4.
  • Available in small laboratory pack sizes (5 g, 10 g, 50 g).
  • Store at room temperature; in solution: -80 °C (2 years), -20 °C (1 year).
  • For research use only; not for sale to patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.