Learn More
Cayman Chemical StearIdonIc AcId 10mg
SDP
Catalog No. 502893406
Encompass_Preferred
Stearidonic acid is an 18-carbon, ω-3 fatty acid which is a dietary precursor to EPA and DHA. Stearidonic acid is present in small amounts in seed oils, especially black currant seed oil.
Catalog No. 50-289-3406 Supplier Cayman Chemical Supplier No. 9032010 MG
Stearidonic acid is an 18-carbon, ω-3 fatty acid which is a dietary precursor to EPA and DHA. Stearidonic acid is present in small amounts in seed oils, especially black currant seed oil.