Learn More
Medchemexpress LLC Stearidonic acid | 20290-75-9 | 98.3% | 276.41 | 10 MG
SDP
Catalog No. 5000437748
Encompass_Preferred
Stearidonic acid is an orally active polyunsaturated fatty acid found in blackcurrant seed oil. It can inhibit 5-lipoxygenase and reduce leukotriene and 5-HETE synthesis, and can increase the content of eicosapentaenoic acid (EPA) in red blood cells. Stearidonic acid can be used for the research of metabolic disease.
  • Inhibits 5-lipoxygenase pathway product formation in isolated human leukocytes.
  • Reduces leukotriene and 5-HETE synthesis.
  • Increases eicosapentaenoic acid (EPA) content in red blood cells.
  • Produces greater enrichment in plasma eicosapentaenoic acid (EPA) in rat models.
Catalog No. 50-004-37748 Supplier Medchemexpress LLC Supplier No. HY11312010MG
Add to Cart
Add to Cart
Stearidonic acid is an orally active polyunsaturated fatty acid found in blackcurrant seed oil. It can inhibit 5-lipoxygenase and reduce leukotriene and 5-HETE synthesis, and can increase the content of eicosapentaenoic acid (EPA) in red blood cells. Stearidonic acid can be used for the research of metabolic disease.
  • Inhibits 5-lipoxygenase pathway product formation in isolated human leukocytes.
  • Reduces leukotriene and 5-HETE synthesis.
  • Increases eicosapentaenoic acid (EPA) content in red blood cells.
  • Produces greater enrichment in plasma eicosapentaenoic acid (EPA) in rat models.