Learn More
Cayman Chemical Stearidonic Acid ethyl ester
SDP
Supplier:  Cayman Chemical 10006856500 UG
Encompass_Preferred
An essential fatty acid in the ω-3 series; an ester of the free acid that is less water soluble, but more amenable for the formulation of stearidonate-containing diets and dietary supplements
Catalog No. 50-288-8756