Learn More
Cayman Chemical Stearidonic Acid ethyl ester, Purity: ≥98%, CAS: 119798-44-6, 10 mg
SDP
Catalog No. 502889451
Encompass_Preferred
An essential fatty acid in the ω-3 series; an ester of the free acid that is less water soluble, but more amenable for the formulation of stearidonate-containing diets and dietary supplements
Catalog No. 50-288-9451 Supplier Cayman Chemical Supplier No. 1000685610 MG
An essential fatty acid in the ω-3 series; an ester of the free acid that is less water soluble, but more amenable for the formulation of stearidonate-containing diets and dietary supplements