Learn More
Cayman Chemical Stearidonic Acid ethyl ester
SDP
Catalog No. 502888752
Encompass_Preferred
An essential fatty acid in the ω-3 series; an ester of the free acid that is less water soluble, but more amenable for the formulation of stearidonate-containing diets and dietary supplements
Catalog No. 50-288-8752 Supplier Cayman Chemical Supplier No. 100068561 MG
Add to Cart
Add to Cart
An essential fatty acid in the ω-3 series; an ester of the free acid that is less water soluble, but more amenable for the formulation of stearidonate-containing diets and dietary supplements