Learn More
Cayman Chemical Stearidonic Acid methyl ester, Purity: ≥98%, CAS: 73097-00-4, 5 mg
SDP
Catalog No. 502890271
Encompass_Preferred
An essential fatty acid in the ω-3 series; an ester of the free acid that is less water soluble, but more amenable for the formulation of stearidonate-containing diets and dietary supplements
Catalog No. 50-289-0271 Supplier Cayman Chemical Supplier No. 100050005 MG
An essential fatty acid in the ω-3 series; an ester of the free acid that is less water soluble, but more amenable for the formulation of stearidonate-containing diets and dietary supplements