Learn More
Cayman Chemical Stearoyl Ethanolamide, Purity: ≥98%, CAS: 111-57-9, 10 mg
SDP
Catalog No. 502895469
Encompass_Preferred
A member of the family of fatty N-acyl ethanolamines collectively called anandamides and the most abundant of several fatty acid ethanolamides produced by the PLD hydrolysis of mouse neuroblastoma cell membrane phospholipids; its specific role in the cannabinergic system remains to be elucidated
Catalog No. 50-289-5469 Supplier Cayman Chemical Supplier No. 9024510 MG
A member of the family of fatty N-acyl ethanolamines collectively called anandamides and the most abundant of several fatty acid ethanolamides produced by the PLD hydrolysis of mouse neuroblastoma cell membrane phospholipids; its specific role in the cannabinergic system remains to be elucidated