Learn More
Medchemexpress LLC Stearoyl glutamic acid sodium | 38517-23-6 | C23H42NNaO5 | 1 G
SDP
Catalog No. 5000355610
Encompass_Preferred
Stearoyl glutamic acid sodium (Sodium stearoyl glutamate) is an anionic emulsifier and surfactant suitable for use in cosmetics. It also functions as an amino acid alkyl amide sensitizer and a chelating agent. While it may cause allergic contact dermatitis, it is noted for its emulsifying properties and preservative efficacy.
  • Increases apparent viscosity in O/W emulsions.
  • Reduces particle size in O/W emulsions.
  • Enhances particle size uniformity in O/W emulsions.
  • Improves long-term physical stability in O/W emulsions.
  • Demonstrates excellent preservative efficacy in FB1 emulsion, eliminating tested bacteria, yeasts, and fungi within 7 days and maintaining microbial sterility for up to 28 days.
Catalog No. 50-003-55610 Supplier Medchemexpress LLC Supplier No. HY1459551G
Stearoyl glutamic acid sodium (Sodium stearoyl glutamate) is an anionic emulsifier and surfactant suitable for use in cosmetics. It also functions as an amino acid alkyl amide sensitizer and a chelating agent. While it may cause allergic contact dermatitis, it is noted for its emulsifying properties and preservative efficacy.
  • Increases apparent viscosity in O/W emulsions.
  • Reduces particle size in O/W emulsions.
  • Enhances particle size uniformity in O/W emulsions.
  • Improves long-term physical stability in O/W emulsions.
  • Demonstrates excellent preservative efficacy in FB1 emulsion, eliminating tested bacteria, yeasts, and fungi within 7 days and maintaining microbial sterility for up to 28 days.