Learn More
Medchemexpress LLC Stearoyl-L-carnitine chloride | 32350-57-5 | 99.9% | 1 MG
SDP
Catalog No. 5000338731
Encompass_Preferred
Stearoyl-L-carnitine chloride is an endogenous long-chain acylcarnitine, acting as a less potent inhibitor of GlyT2. It inhibits glycine responses by 16.8% at concentrations up to 3 μM.
  • Inhibits glycine transport by GlyT2.
  • Used in research for Alzheimer's disease, mild cognitive impairment, and subjective memory complaint.
  • Soluble in ethanol and DMSO for various in vitro applications.
  • Solid storage at -20°C, sealed and away from moisture.
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-003-38731 Supplier Medchemexpress LLC Supplier No. HY130466S1MG
Stearoyl-L-carnitine chloride is an endogenous long-chain acylcarnitine, acting as a less potent inhibitor of GlyT2. It inhibits glycine responses by 16.8% at concentrations up to 3 μM.
  • Inhibits glycine transport by GlyT2.
  • Used in research for Alzheimer's disease, mild cognitive impairment, and subjective memory complaint.
  • Soluble in ethanol and DMSO for various in vitro applications.
  • Solid storage at -20°C, sealed and away from moisture.
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month.