Learn More
Medchemexpress LLC Stearoyl-L-carnitine chloride | 32350-57-5 | 99.9% | 100 MG
SDP
Catalog No. 5000339958
Encompass_Preferred
Stearoyl-L-carnitine chloride is an endogenous long-chain acylcarnitine. It acts as a less potent inhibitor of GlyT2 and inhibits glycine responses by 16.8% at concentrations up to 3 μM. This compound is significantly decreased in Alzheimer's disease (AD), mild cognitive impairment (MCI), and subjective memory complaint (SMC), suggesting its potential role in neurodegenerative research.
  • Endogenous long-chain acylcarnitine
  • Acts as a less potent inhibitor of GlyT2
  • Inhibits glycine responses by 16.8% at concentrations up to 3 μM
  • Inhibits Na+-dependent [³H]carnitine uptake by approximately 50% in HPCT cells
  • Associated with neurological conditions such as Alzheimer's disease
Catalog No. 50-003-39958 Supplier Medchemexpress LLC Supplier No. HY130466100MG
Stearoyl-L-carnitine chloride is an endogenous long-chain acylcarnitine. It acts as a less potent inhibitor of GlyT2 and inhibits glycine responses by 16.8% at concentrations up to 3 μM. This compound is significantly decreased in Alzheimer's disease (AD), mild cognitive impairment (MCI), and subjective memory complaint (SMC), suggesting its potential role in neurodegenerative research.
  • Endogenous long-chain acylcarnitine
  • Acts as a less potent inhibitor of GlyT2
  • Inhibits glycine responses by 16.8% at concentrations up to 3 μM
  • Inhibits Na+-dependent [³H]carnitine uptake by approximately 50% in HPCT cells
  • Associated with neurological conditions such as Alzheimer's disease