Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stearoyl-L-carnitine-d3 chloride | 2245711-27-5 | MFCD08705119 | 99.8% | C25H47D3ClNO4 | 10MG
SDP

Catalog No. 5000260665
Encompass_Preferred

Stearoyl-L-carnitine-d3 chloride is a deuterium-labeled analogue of stearoyl-L-carnitine chloride supplied as an isotope-labeled internal standard for analytical and metabolic research. The product is intended for use in quantitative mass spectrometry and metabolite profiling where a stable isotope reference improves accuracy and reproducibility. It is provided as a high-purity solid with recommended storage conditions to maintain stability.

  • Deuterium-labeled internal standard for accurate quantification.
  • High purity (99.8%) for reliable analytical performance.
  • Chemical formula C25H47D3ClNO4; deuterium (d3) labeled.
  • Suitable for LC/MS and GC/MS applications.
  • Stable when stored sealed; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied in small research sizes suitable for method development and calibration.

Catalog No. 50-002-60665 Supplier Medchemexpress LLC Supplier No. HY130466S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stearoyl-L-carnitine-d3 chloride is a deuterium-labeled analogue of stearoyl-L-carnitine chloride supplied as an isotope-labeled internal standard for analytical and metabolic research. The product is intended for use in quantitative mass spectrometry and metabolite profiling where a stable isotope reference improves accuracy and reproducibility. It is provided as a high-purity solid with recommended storage conditions to maintain stability.

  • Deuterium-labeled internal standard for accurate quantification.
  • High purity (99.8%) for reliable analytical performance.
  • Chemical formula C25H47D3ClNO4; deuterium (d3) labeled.
  • Suitable for LC/MS and GC/MS applications.
  • Stable when stored sealed; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied in small research sizes suitable for method development and calibration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.