Learn More
Stearoyl-L-carnitine-(N-methyl-d3), ≥97.0% (TLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 1814920-07-4 |
| Linear Formula | C25D3H46 NO4 |
| Synonym | (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxooctadecyl)oxy]-1-propanaminium inner salt; L-Carnitine-(methyl-d3) octadecanoyl ester; Octadecanoyl-L-carnitine-(N-methyl-d3) |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 430.68 |
| Percent Purity | ≥97.0% (TLC) |
| Grade | Analytical Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.