Learn More
Cayman Chemical Stearoyl Serotonin, Purity: ≥95%, CAS: 67964-87-8, 10 mg
SDP
Catalog No. 502893328
Encompass_Preferred
A hybrid molecule patterned after arachidonoyl serotonin; while saturated 11- or 12-fatty acids linked to serotonin potently inhibit capsaicin-induced TRPV1 channel activation (IC50 =0.76 μM) without blocking FAAH-mediated hydrolysis of arachidonoyl ethanolamine (IC50 > 50 μM), the effects of stearoyl serotonin are unknown
Catalog No. 50-289-3328 Supplier Cayman Chemical Supplier No. 900063110 MG
A hybrid molecule patterned after arachidonoyl serotonin; while saturated 11- or 12-fatty acids linked to serotonin potently inhibit capsaicin-induced TRPV1 channel activation (IC50 =0.76 μM) without blocking FAAH-mediated hydrolysis of arachidonoyl ethanolamine (IC50 > 50 μM), the effects of stearoyl serotonin are unknown