Learn More
Medchemexpress LLC Stearoylcarnitine | 25597-09-5 | 99.9% | 427.66 | 10 MG
SDP
Catalog No. 5000437241
Encompass_Preferred
Stearoylcarnitine is a fatty ester lipid molecule and an endogenous metabolite. It can function as a PKC inhibitor and accumulates in β cells, contributing to impaired insulin synthesis and energy deficiency in type 2 diabetes mouse models. Additionally, Stearoylcarnitine inhibits lecithin cholesterol acyltransferase (LCAT) in rat and rabbit plasma and serves as a metabolomics biomarker for Parkinson's disease. It is also noted as a less potent inhibitor of GlyT2.
  • Endogenous metabolite.
  • Can be used as a PKC inhibitor.
  • Accumulates in β cells, leading to arrest of insulin synthesis and energy deficiency in type 2 diabetes mouse.
  • Inhibits lecithin cholesterol acyltransferase (LCAT) in rat and rabbit plasma.
  • Acts as a metabolomics biomarker for Parkinson's disease.
  • Less potent inhibitor of GlyT2.
Catalog No. 50-004-37241 Supplier Medchemexpress LLC Supplier No. HY11320210MG
Stearoylcarnitine is a fatty ester lipid molecule and an endogenous metabolite. It can function as a PKC inhibitor and accumulates in β cells, contributing to impaired insulin synthesis and energy deficiency in type 2 diabetes mouse models. Additionally, Stearoylcarnitine inhibits lecithin cholesterol acyltransferase (LCAT) in rat and rabbit plasma and serves as a metabolomics biomarker for Parkinson's disease. It is also noted as a less potent inhibitor of GlyT2.
  • Endogenous metabolite.
  • Can be used as a PKC inhibitor.
  • Accumulates in β cells, leading to arrest of insulin synthesis and energy deficiency in type 2 diabetes mouse.
  • Inhibits lecithin cholesterol acyltransferase (LCAT) in rat and rabbit plasma.
  • Acts as a metabolomics biomarker for Parkinson's disease.
  • Less potent inhibitor of GlyT2.