Learn More
Medchemexpress LLC Stearoylcarnitine | 25597-09-5 | 99.9% | 427.66 | 50 MG
SDP
Catalog No. 5000437864
Encompass_Preferred
Stearoylcarnitine is a fatty ester lipid molecule and an endogenous metabolite. It can act as a PKC inhibitor and accumulates in β cells, leading to arrest of insulin synthesis and energy deficiency in type 2 diabetes mouse models. It also inhibits lecithin cholesterol acyltransferase (LCAT) in rat and rabbit plasma and serves as a metabolomics biomarker for Parkinson's disease. Additionally, it is a less potent inhibitor of GlyT2.
  • Functions as an endogenous metabolite.
  • Acts as a PKC inhibitor.
  • Accumulates in beta cells, impairing insulin synthesis.
  • Inhibits lecithin cholesterol acyltransferase (LCAT) in rat and rabbit plasma.
  • Serves as a metabolomics biomarker for Parkinson's disease.
  • Less potent inhibitor of GlyT2.
  • Impairs mitochondrial energy metabolism in MIN6 cells.
  • Alters brain acylcarnitine levels in a mouse model of hypoxic-ischemic brain injury.
Catalog No. 50-004-37864 Supplier Medchemexpress LLC Supplier No. HY11320250MG
Stearoylcarnitine is a fatty ester lipid molecule and an endogenous metabolite. It can act as a PKC inhibitor and accumulates in β cells, leading to arrest of insulin synthesis and energy deficiency in type 2 diabetes mouse models. It also inhibits lecithin cholesterol acyltransferase (LCAT) in rat and rabbit plasma and serves as a metabolomics biomarker for Parkinson's disease. Additionally, it is a less potent inhibitor of GlyT2.
  • Functions as an endogenous metabolite.
  • Acts as a PKC inhibitor.
  • Accumulates in beta cells, impairing insulin synthesis.
  • Inhibits lecithin cholesterol acyltransferase (LCAT) in rat and rabbit plasma.
  • Serves as a metabolomics biomarker for Parkinson's disease.
  • Less potent inhibitor of GlyT2.
  • Impairs mitochondrial energy metabolism in MIN6 cells.
  • Alters brain acylcarnitine levels in a mouse model of hypoxic-ischemic brain injury.