Learn More
Cayman Chemical Stearoyl-L-carnitine (chloride), Purity: ≥95%, CAS: 32350-57-5, 50 mg
SDP
Catalog No. 502934825
Encompass_Preferred
A long-chain acylcarnitine; inhibits sodium-dependent [3H]carnitine uptake in HPCT cells by 52% at 500 μM; inhibits lecithin:cholesterol acyltransferase activity in rat, but not human, plasma at 500 μM/ml of plasma; levels are decreased in plasma of patients with chronic fatigue syndrome and increased in patients with end-stage renal disease
Catalog No. 50-293-4825 Supplier Cayman Chemical Supplier No. 2655650 MG
A long-chain acylcarnitine; inhibits sodium-dependent [3H]carnitine uptake in HPCT cells by 52% at 500 μM; inhibits lecithin:cholesterol acyltransferase activity in rat, but not human, plasma at 500 μM/ml of plasma; levels are decreased in plasma of patients with chronic fatigue syndrome and increased in patients with end-stage renal disease