Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stearyl gallate | 10361-12-3 | 10 G
SDP

Catalog No. 5000417313
Encompass_Preferred

Stearyl gallate is an alkyl gallate characterized by a long alkyl chain (18 carbons). It demonstrates both antioxidant activity and weak antiviral properties, specifically against HSV-1.

  • Exhibits antioxidant activity
  • Shows weak antiviral activity against HSV-1
  • Has a purity of 99.50%
  • Appears as a white to off-white solid
  • Stable under recommended storage conditions
  • Melting point of 95°C
  • Soluble in DMSO

Catalog No. 50-004-17313 Supplier Medchemexpress LLC Supplier No. HYN808210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stearyl gallate is an alkyl gallate characterized by a long alkyl chain (18 carbons). It demonstrates both antioxidant activity and weak antiviral properties, specifically against HSV-1.

  • Exhibits antioxidant activity
  • Shows weak antiviral activity against HSV-1
  • Has a purity of 99.50%
  • Appears as a white to off-white solid
  • Stable under recommended storage conditions
  • Melting point of 95°C
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.