Learn More
Medchemexpress LLC Stearyl gallate | 10361-12-3 | 50 G
SDP
Catalog No. 5000417689
Encompass_Preferred
Stearyl gallate is an alkyl gallate with a long alkyl chain (18 carbon atoms). It exhibits antioxidant activity and a weak antiviral activity against HSV-1. It is for research use only and not sold to patients. The purity of the product is 99.50%.
  • Alkyl gallate with a long alkyl chain (18 carbon atoms)
  • Exhibits antioxidant activity
  • Weak antiviral activity against HSV-1
  • For research use only
  • Purity of 99.50%
Catalog No. 50-004-17689 Supplier Medchemexpress LLC Supplier No. HYN808250G
Stearyl gallate is an alkyl gallate with a long alkyl chain (18 carbon atoms). It exhibits antioxidant activity and a weak antiviral activity against HSV-1. It is for research use only and not sold to patients. The purity of the product is 99.50%.
  • Alkyl gallate with a long alkyl chain (18 carbon atoms)
  • Exhibits antioxidant activity
  • Weak antiviral activity against HSV-1
  • For research use only
  • Purity of 99.50%