Learn More
Medchemexpress LLC Stearyldiethanolamine | 10213-78-2 | 100 MG
SDP
Catalog No. 5000339596
Encompass_Preferred
Stearyldiethanolamine is an amine-based surfactant used in the development of antibacterial freshness-keeping film or antibacterial nonwoven fabric. It can also serve as a collector for quartz and malachite surfaces, efficiently recovering malachite. This compound disrupts bacterial cell membranes with its alkyl chain, leading to irreversible damage and cell death.
  • Used in antibacterial freshness-keeping film.
  • Used in antibacterial nonwoven fabric.
  • Functions as a collector for quartz and malachite.
  • Efficiently recovers malachite from quartz.
  • Disrupts bacterial cell membranes.
  • Increases recovery of malachite and quartz dose-dependently.
  • Exhibits inhibitory effects against Gram-positive and Gram-negative bacteria.
Catalog No. 50-003-39596 Supplier Medchemexpress LLC Supplier No. HY129197100MG
Stearyldiethanolamine is an amine-based surfactant used in the development of antibacterial freshness-keeping film or antibacterial nonwoven fabric. It can also serve as a collector for quartz and malachite surfaces, efficiently recovering malachite. This compound disrupts bacterial cell membranes with its alkyl chain, leading to irreversible damage and cell death.
  • Used in antibacterial freshness-keeping film.
  • Used in antibacterial nonwoven fabric.
  • Functions as a collector for quartz and malachite.
  • Efficiently recovers malachite from quartz.
  • Disrupts bacterial cell membranes.
  • Increases recovery of malachite and quartz dose-dependently.
  • Exhibits inhibitory effects against Gram-positive and Gram-negative bacteria.